missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Cat IgG F(ab')2 fragment, AP, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10472.1mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Cat IgG F(ab')2 fragment | |
| Polyclonal | |
| Goat anti-cat IgG recognizes cat IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase. | |
| In 50mM Tris-Cl, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl2, 50% (v/v) glycerol, pH 8.0 (0.01% sodium azide) | |
| Cat IgG F(ab')2 fragment | |
| 1 mg | |
| Feline | |
| Liquid |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| Alkaline Phosphatase (AP) | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C. Do not Freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction