missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carcino Embryonic Antigen, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native Carcino Embryonic Antigen.
Brand: Abnova PAB7939.1mg
This item is not returnable.
View return policy
Specifications
| Carcino Embryonic Antigen | |
| Polyclonal | |
| Goat polyclonal antibody raised against native Carcino Embryonic Antigen. | |
| In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) | |
| Native purified human Carcino Embryonic Antigen. | |
| RUO | |
| This antibody is specific to human CEA. No cross-reactivity with human NCA, NCA2 or BGP1 (Biliary glycoprotein-l). | |
| Store at 4°C. |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction