missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG Fc Secondary Antibody, Rhodamine, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73349-1MG
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal specifically detects IgG Fc in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgGFc | |
| Polyclonal | |
| Rhodamine | |
| Lyophilized from 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, with 10Mg/ml Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| Dog IgG Fc fragment | |
| 1 mg | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Dog IgG, Dog IgG F(c) and Dog Serum. No reaction was observed against Dog IgG F(ab). | |
| Canine | |
| IgG |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Goat | |
| Affinity Purified | |
| Secondary | |
| Reconstitute with 1.0mL of deionized water. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction