missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgE Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7343B
This item is not returnable.
View return policy
Description
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Specifications
| IgE Heavy Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Canine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot | |
| Biotin | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Dog IgE | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction