missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgA Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7308PCP
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| PBS | |
| Dog IgA | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PerCP | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction