Learn More
CACNA1C, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CACNA1C.
Brand: Abnova PAB11506.100ug
Description
This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by this gene binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins. [provided by RefSeq
Sequence: C-RARGRPSEEELQDSpécification
| CACNA1C | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CACNA1C | |
| CACH2/CACN2/CACNL1A1/CCHL1A1/CaV1.2/MGC120730/TS | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 775 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CACNA1C. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000710.5. | |
| CACNA1C | |
| A synthetic peptide corresponding to human CACNA1C. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only