Learn More
C1D, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of C1D.
Brand: Abnova PAB11451.100ug
Description
The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Several alternatively spliced transcript variants of this gene have been described, but the full length nature of some of these variants has not been determined. [provided by RefSeq
Sequence: C-SKNASKVANKGKSKSSpecifications
| C1D | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| C1D | |
| MGC12261/MGC14659/SUNCOR | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10438 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of C1D. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_775269.1 | |
| C1D | |
| A synthetic peptide corresponding to human C1D. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only