Learn More
BTD, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BTD.
Brand: Abnova PAB27622.100ug
Description
Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency. [provided by RefSeq
Sequence: EKDAQEVHCDEATKSpecifications
| BTD | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (0.5-1.5ug/ml) The optimal working dilution should be determined by the end user. | |
| BTD | |
| BTD | |
| A synthetic peptide corresponding to amino acids 372-385 at internal region of human BTD. | |
| 100 μg | |
| Primary | |
| Human |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of BTD. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_000051.1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 686 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only