Learn More
BRF2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BRF2.
Brand: Abnova PAB6262.100ug
Description
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein. [provided by RefSeq
Sequence: C-QAARQAATSVPNPPSpecifications
| BRF2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRF2 | |
| BRFU/FLJ11052/TFIIIB50 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 55290 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of BRF2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_060780 | |
| BRF2 | |
| A synthetic peptide corresponding to human BRF2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only