missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Galactosidase, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native Beta-Galactosidase.
Brand: Abnova PAB15374.1mL
This item is not returnable.
View return policy
Specifications
| Beta-Galactosidase | |
| Polyclonal | |
| Goat polyclonal antibody raised against native Beta-Galactosidase. | |
| In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 | |
| Native purified Escherichia coli Galactosidase-beta. | |
| RUO | |
| This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques. This product is specially prepared for the detection and purification of recombinant proteins. It shows reactivity to beta-galactosidase and is tested by Western or dot blot to ensure monospecificity using lysates of D-lac and strain 3.3 of E. coli. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunodiffusion, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:500) ELISA (1:400-1:2000) The optimal working dilution should be determined by the end user. | |
| Goat | |
| 1 mL | |
| Primary | |
| E. coli | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction