missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAZ2B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BAZ2B.
Brand: Abnova PAB6312.100ug
This item is not returnable.
View return policy
Description
Sequence: C-YFEKKWTDTFKVSSpecifications
| BAZ2B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| BAZ2B | |
| DKFZp434H071/DKFZp762I0516/FLJ45644/WALp4 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 29994 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of BAZ2B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_038478 | |
| BAZ2B | |
| A synthetic peptide corresponding to human BAZ2B. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction