Learn More
B2M Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of B2M.
Brand: Abnova PAB25444.100ug
Description
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia
Sequence: QRTPKIQVYSRHPCSpecifications
| B2M | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| B2M | |
| B2M | |
| A synthetic peptide corresponding to N-terminus of human B2M. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of B2M. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_004039.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 567 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only