Learn More
AXIN1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of AXIN1.
Brand: Abnova PAB6140.100ug
Description
This gene encodes a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a dishevelled and axin (DIX) domain. The encoded protein interacts with adenomatosis polyposis coli, catenin (cadherin-associated protein), beta 1, 88kDa, glycogen synthase kinase 3 beta, protein phosphate 2, and itself. This protein functions as a negative regulator of the wingless-type MMTV integration site family, member 1 (WNT) signaling pathway and can induce apoptosis. The crystal structure of a portion of this protein, alone and in a complex with other proteins, has been resolved. Mutations in this gene have been associated with hepatocellular carcinoma, hepatoblastomas, ovarian endometriod adenocarcinomas, and medullablastomas. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: C-FEEKIIGKVEKVDSpecifications
| AXIN1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| AXIN1 | |
| AXIN/MGC52315 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8312 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of AXIN1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_003493.1;NP_851393.1 | |
| AXIN1 | |
| A synthetic peptide corresponding to human AXIN1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only