Learn More
ARSD, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ARSD.
Brand: Abnova PAB11496.100ug
Description
The protein encoded by this gene is a member of the sulfatase family. Sulfatases are essential for the correct composition of bone and cartilage matrix. This encoded protein is postranslationally glycosylated and localized to the lysosome. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-DASNGYRALQWNASpecifications
| ARSD | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARSD | |
| ARSD | |
| A synthetic peptide corresponding to human ARSD. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ARSD. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001660.2;NP_033667.2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 414 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only