Learn More
ARPC2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ARPC2.
Brand: Abnova PAB6168.100ug
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined. [provided by RefSeq
Sequence: C-EMKTITGKTFSSRSpecifications
| ARPC2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western blot (0.1-0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARPC2 | |
| ARC34/PNAS-139/PRO2446/p34-Arc | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10109 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ARPC2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005722;NP_690601 | |
| ARPC2 | |
| A synthetic peptide corresponding to human ARPC2. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms, as represented by NP_005722 and NP_690601. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only