Learn More
ARPC1B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ARPC1B.
Brand: Abnova PAB6170.100ug
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. [provided by RefSeq
Sequence: AYHSFLVEPISCHSpecifications
| ARPC1B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARPC1B | |
| ARC41/p40-ARC/p41-ARC | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10095 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ARPC1B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005711 | |
| ARPC1B | |
| A synthetic peptide corresponding to human ARPC1B. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only