missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Armenian Hamster IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6887
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Armenian Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Armenian Hamster IgG whole molecule | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| Minimal cross reactivity is expected against golden Syrian hamster IgG. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, ChIP Assay | |
| Unconjugated | |
| Goat | |
| Delipidation and Defibrination | |
| RUO | |
| Secondary | |
| Armenian Hamster | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction