missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Armenian Hamster IgG (H+L) Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA129558
This item is not returnable.
View return policy
Description
PA1-29558 detects Hamster (Armenian) IgG from hamster samples.
Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Armenian Hamster IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Hamster | |
| Secondary Antibody | |
| IgG |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol, 1mM MgCl2, 0.1mM ZnCl2 and 0.1% sodium azide; pH 8 | |
| Armenian Hamster IgG whole molecule. | |
| 500 μg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction