Learn More
AR, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of AR.
Brand: Abnova PAB6663.100ug
Description
The androgen receptor gene is more than 90kb long and codes for a protein that has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Two alternatively spliced variants encoding distinct isoforms have been described. [provided by RefSeq
Sequence: EVQLGLGRVYPRPPSCSpecifications
| AR | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| AR | |
| AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 367 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of AR. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000035 | |
| AR | |
| A synthetic peptide corresponding to human AR. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only