Learn More
APOL3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of APOL3.
Brand: Abnova PAB11448.100ug
Description
This gene is a member of the apolipoprotein L gene family, and it is present in a cluster with other family members on chromosome 6. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids, including cholesterol, and/or allow the binding of lipids to organelles. In addition, expression of this gene is up-regulated by tumor necrosis factor-alpha in endothelial cells lining the normal and atherosclerotic iliac artery and aorta. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: ELTQIYQRLNPCHTHSpecifications
| APOL3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| APOL3 | |
| APOLIII/CG12-1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 80833 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of APOL3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_663615.1;NP_055164.1;NP_663616.1 | |
| APOL3 | |
| A synthetic peptide corresponding to human APOL3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only