Learn More
ANTXR1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ANTXR1.
Brand: Abnova PAB6677.100ug
Description
This gene encodes a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colorectal cancer. The encoded protein has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax. The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to this receptor protein mediates delivery of toxin components to the cytosol of cells. Once inside the cell, the other two components of anthrax toxin, edema factor (EF) and lethal factor (LF) disrupt normal cellular processes. Three alternatively spliced variants that encode different protein isoforms have been described. [provided by RefSeq
Sequence: C-RAPPPSRPPPRPSVSpecifications
| ANTXR1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANTXR1 | |
| ATR/FLJ10601/FLJ11298/FLJ21776/TEM8 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84168 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ANTXR1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_115584 | |
| ANTXR1 | |
| A synthetic peptide corresponding to human ANTXR1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize variant 1 (NP_115584) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only