Learn More
ANGPT1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ANGPT1.
Brand: Abnova PAB19687.100ug
Description
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart. [provided by RefSeq
Sequence: C-PDFSSQKLQHSpecifications
| ANGPT1 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of ANGPT1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001137.2 | |
| ANGPT1 | |
| A synthetic peptide corresponding to internal region of human ANGPT1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| ANGPT1 | |
| AGP1/AGPT/ANG1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 284 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only