Learn More
ACTR1A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ACTR1A.
Brand: Abnova PAB6617.100ug
Description
This gene encodes a 42.6kDa subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin. [provided by RefSeq
Sequence: C-YEEDGARSIHRKTSpecifications
| ACTR1A | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACTR1A | |
| ARP1/CTRN1/FLJ52695/FLJ52800/FLJ55002 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10121 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ACTR1A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005727.1 | |
| ACTR1A | |
| A synthetic peptide corresponding to human ACTR1A. | |
| 100 μg | |
| Primary | |
| This epitope was selected to minimise the chance of cross-reactivity with ACTR1B (YEEDGSRAIHRKT). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only