missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL6A/ACTL6B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ACTL6A/ACTL6B.
Brand: Abnova PAB6615.100ug
This item is not returnable.
View return policy
Description
Sequence: YEEGGKQCVERKCPSpecifications
| ACTL6A/ACTL6B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| NP_057272 (Gene ID : 51412);NP_004292 (Gene ID : 86) | |
| ACTL6B | |
| A synthetic peptide corresponding to human ACTL6A/ACTL6B. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both BAF53A and BAF53B, which are almost identical. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ACTL6A/ACTL6B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ACTL6/BAF53B | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 51412/86 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction