Learn More
ACSL4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ACSL4.
Brand: Abnova PAB6260.100ug
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme preferentially utilizes arachidonate as substrate. The absence of this enzyme may contribute to the mental retardation or Alport syndrome. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq
Sequence: C-HYLKDIERMYGGKSpecifications
| ACSL4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACSL4 | |
| ACS4/FACL4/LACS4/MRX63/MRX68 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2182 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ACSL4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_004449;NP_075266 | |
| ACSL4 | |
| A synthetic peptide corresponding to human ACSL4. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported human isoforms, as represented by NP_004449 and NP_075266. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only