missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD14B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ABHD14B.
Brand: Abnova PAB25440.100ug
This item is not returnable.
View return policy
Description
Sequence: HPCYLDKPEEWHTSpecifications
| ABHD14B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ABHD14B | |
| CIB/MGC15429 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84836 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ABHD14B. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_116139.1 | |
| ABHD14B | |
| A synthetic peptide corresponding to amino acids 188-200 at internal region of human ABHD14B. | |
| 100 μg | |
| Primary | |
| Reported variants represent identical protein: NP_116139.1, NP_001139786.1. This antibody is expected to NOT cross-react with ABHD14A. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction