missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GO Protein alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38002-20ul
This item is not returnable.
View return policy
Description
GO Protein alpha Polyclonal antibody specifically detects GO Protein alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GO Protein alpha | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| DKFZp686O0962, G-ALPHA-o, GNAO, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide O, guanine nucleotide binding protein, alpha activating polypeptide O, guanine nucleotide-binding protein G(o) subunit alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human GO Protein alpha (NP_066268.1).,, Sequence:, GFSGEDVKQYKPVVYSNTIQSLAAIVRAMDTLGIEYGDKERKADAKMVCDVVSRMEDTEPFSAELLSAMMRLWGDSGIQECFNRSREYQLNDSAKYYLDSLDRIGAADYQPTEQDILRTRV | |
| 20 μL | |
| Signal Transduction | |
| 2775 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction