missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Go 6983, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 3-[1-[3-(Dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione |
| CAS | 133053-19-7 |
| Molecular Formula | C26H26N4O3 |
| Quantity | 10 mg |
| Synonym | Go 6983, Goe 6983 |
| InChI Formula | InChI=1S/C26H26N4O3/c1-29(2)11-6-12-30-15-20(18-13-16(33-3)9-10-22(18)30)24-23(25(31)28-26(24)32)19-14-27-21-8-5-4-7-17(19)21/h4-5,7-10,13-15,27H,6,11-12H2,1-3H3,(H,28,31,32) |
| SMILES | COc1ccc2c(c1)c(cn2CCCN(C)C)C3=C(C(=O)NC3=O)c4c[nH]c5ccccc45 |
| IUPAC Name | 3-[1-[3-(dimethylamino)propyl]-5-methoxyindol-3-yl]-4-(1H-indol-3-yl)pyrrole-2,5-dione |
| Molecular Weight (g/mol) | 442.51 |
| Formula Weight | 442.2005 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction