missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRHR Rabbit anti-Human, Clone: GNRHR/2982R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | GnRHR |
|---|---|
| Clone | GNRHR/2982R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
GnRHR Monoclonal specifically detects GnRHR in Human samples. It is validated for ELISA.Specifications
| GnRHR | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
| Recombinant full-length human GnRHR protein | |
| Primary | |
| Store at 4°C. |
| GNRHR/2982R | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 2798 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title