missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNRHR, Mouse anti-Human, Clone: AT2.G7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB8771.100ug
This item is not returnable.
View return policy
Specifications
| GNRHR | |
| Monoclonal | |
| 1 μg/μL | |
| Western Blot (1:1000) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user. | |
| gonadotropin-releasing hormone receptor | |
| GNRHR | |
| A mixture of peptides from different regions of human GNRHR. | |
| RUO | |
| 2798 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| AT2.G7 | |
| Unconjugated | |
| In PBS, pH 7.4 (0.05% sodium azide) | |
| GNRHR1/GRHR/LHRHR/LRHR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction