missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GnRHR Antibody (LCHR37), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27924
This item is not returnable.
View return policy
Description
GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| GnRHR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL | |
| P30968 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| LCHR37 | |
| Unconjugated | |
| PBS, 0.1 mg/mL BSA | |
| GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
| Recombinant full-length human protein was used as the immunogen for the GnRHR antibody. | |
| 100 μg | |
| GPCR | |
| 2798 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction