missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRHR Antibody (GNRHR/2982R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08770PE
This item is not returnable.
View return policy
Description
GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for ELISA
Specifications
| GnRHR | |
| Monoclonal | |
| PE | |
| GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
| Recombinant full-length human GnRHR protein | |
| 0.1 mL | |
| GPCR | |
| 2798 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| GNRHR/2982R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction