missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GnRH2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552076

Product Code. 15730031

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HWSHGWYPGG KRALSSAQDP QNALRPPAGS PVQTAHGLPS DALAPLDDSM PWEGRTTAQW SLHRKRHLAR TLLTAAREPR Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 30%.

This gene encodes a secreted peptide hormone and member of the gonadotropin-releasing hormone (GnRH) family of proteins. The encoded protein regulates reproductive function by stimulating the production and release of the gonadotropins follicle-stimulating hormone (FSH) and luteinizing hormone (LH). The encoded protein may inhibit endometrial, ovarian, prostate, and breast cancer cell proliferation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GnRH2
Polyclonal
Unconjugated
GNRH2
GnRH II; GNRH2; GnRH-associated peptide 2; GnRH-associated peptide II; GnRH-II; Gonadoliberin II; Gonadoliberin-2; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 2; Gonadotropin-releasing hormone II; LH-RH II; LH-RHII; luliberin II; Luteinizing hormone-releasing hormone II; progonadoliberin II; Progonadoliberin-2
Rabbit
Antigen affinity chromatography
RUO
2797
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43555
GNRH2
Recombinant protein corresponding to Human GnRH2. Recombinant protein control fragment (Product #RP-101638).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.