missing translation for 'onlineSavingsMsg'
Learn More

GnRH1 Polyclonal Antibody, Invitrogen™

Product Code. 11575922
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11575922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11575922 Supplier Invitrogen Supplier No. PA1120

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-120 detects gonadotropin releasing hormone (GnRH) in mouse brain. PA1-120 has been successfully used in immunofluorescence and immunohistochemistry procedures. Immunofluorescent staining of GnRH in mouse embryo brain tissue using PA1-120 results in staining of neurons in the rostral forebrain. In immunohistochemical procedures, 4% paraformaldehyde is recommended as a fixative. The PA1-120 immunogen is a synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. This peptide (Cat. # PEP-168) is available for use in neutralization and control experiments.

GnRH1 Polyclonal antibody specifically detects GnRH1 in Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GnRH1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GNRH1
Gene Accession No. P13562
Gene Alias Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Gene Symbols GNRH1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14714
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.