missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRH-Receptor / LH-RH Receptor Mouse, Clone: GNRHR/768, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2798-MSM3-P1
This item is not returnable.
View return policy
Specifications
| GnRH-Receptor / LH-RH Receptor | |
| Cytoplasmic | |
| GNRHR/768 | |
| Unconjugated | |
| NP_000224 | |
| Mouse | |
| 54-60kDa | |
| Protein A/G Purified | |
| RUO | |
| 3973 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| GNRHR, LHCGR | |
| Recombinant full-length human GNRHR protein | |
| T47D cells. Pituitary gland, ovarian or breast cancers. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction