missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNPTAB Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | GNPTAB |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18350996
|
Novus Biologicals
NBP3-05029-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362204
|
Novus Biologicals
NBP3-05029-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GNPTAB Polyclonal specifically detects GNPTAB in Human samples. It is validated for Western Blot.Specifications
| GNPTAB | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 79158 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp762B226, GlcNAc phosphotransferase, glucosamine (UDP-N-acetyl)-lysosomal-enzyme N-acetylglucosamine phosphotransferase, GNPTA, ICD, KIAA1208, MGC4170, N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits, stealth protein GNPTAB, UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosamine | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-350 of human GNPTAB (NP_077288.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title