missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNPTAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569636
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Catalyzes the formation of mannose 6-phosphate (M6P) markers on high mannose type oligosaccharides in the Golgi apparatus. M6P residues are required to bind to the M6P receptors (MPR), which mediate the vesicular transport of lysosomal enzymes to the endosomal/prelysosomal compartment.
Specifications
| GNPTAB | |
| Polyclonal | |
| Unconjugated | |
| GNPTAB | |
| EG432486; GlcNAc phosphotransferase; glcNAc-1-phosphotransferase subunits alpha/beta; glucosamine (UDP-N-acetyl)-lysosomal-enzyme N-acetylglucosamine phosphotransferase; Gnpta; gnptab; ICD; KIAA1208; mKIAA1208; N-acetylglucosamine-1-phosphate transferase alpha and beta subunits; N-acetylglucosamine-1-phosphate transferase subunits alpha and beta; N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits; N-acetylglucosamine-1-phosphotransferase subunit alpha; N-acetylglucosamine-1-phosphotransferase subunit beta; N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; RGD1564821; si:ch211-234f20.3; Stealth protein gnptab; UDP-N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosamine; Unknown (protein for MGC:122985); wu:fd55d09; zgc:122985; zgc:122985 protein; zgc:158502 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3T906 | |
| GNPTAB | |
| synthetic peptide directed towards the N terminal region of human GNPTAB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction