missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNPNAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113303
This item is not returnable.
View return policy
Description
GNPNAT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).
Specifications
| GNPNAT1 | |
| Polyclonal | |
| Unconjugated | |
| GNPNAT1 | |
| AU017428; AU040593; EMeg32; Glucosamine 6-phosphate N-acetyltransferase; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase 1; glucosamine-phosphate N-acetyltransferase 1; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase; GNA1; GNPNAT; GNPNAT1; Gpnat1; Gsnpat; Phosphoglucosamine acetylase; phosphoglucosamine transacetylase; Protein EMeg32; RGD1563144 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54342, 64841 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96EK6, Q9JK38 | |
| GNPNAT1 | |
| Recombinant Human Glucosamine 6-phosphate N-acetyltransferase protein (1-184AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur