missing translation for 'onlineSavingsMsg'
Learn More

GNPDA2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18411973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411973 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411973 Supplier Novus Biologicals Supplier No. NBP176997AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GNPDA2 Polyclonal antibody specifically detects GNPDA2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNPDA2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.99.6, GlcN6P deaminase 2, glucosamine-6-phosphate deaminase 24921523I18Rik, glucosamine-6-phosphate isomerase 2, Glucosamine-6-phosphate isomerase SB52, GNP2SB52, GNPDA 2, putative glucosamine-6-phosphate isomerase
Host Species Rabbit
Immunogen Antibody was raised against an 8 amino acid synthetic peptide near the carboxy terminus of human GNPDA2. The immunogen is located within the last 50 amino acids of GNPDA2. Amino Acid Squence: ASAGLFSMKDGN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 132789
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.