missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNPDA1 Monoclonal Antibody (237CT2.5.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537517
This item is not returnable.
View return policy
Description
GNPDA1 Monoclonal Antibody for Western Blot
GNPDA1 is a 289 amino acid cytoplasmic protein belonging to the glucosamine/ galactosamine-6-phosphate isomerase family. It is one of the two mammalian glucosamine-6-phosphate deaminase enzymes. GNPDA1 is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium with an aldo/keto isomerization and an amination/deamination. GNPDA1 may trigger calcium oscillations in mammalian eggs at fertilization which in turn serve as an important factor for egg development and early embryo development. It is a homohexamer, ubiquitously expressed in most of human tissues with high expression in testis, ovary, placenta, and heart and also in various cell lines.
Specifications
| GNPDA1 | |
| Monoclonal | |
| Unconjugated | |
| GNPDA1 | |
| glcN6P deaminase 1; glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; Gnp1; GNPDA; GNPDA 1; GNPDA1; Gnpi; GPI; HLN; hypothetical protein LOC550565; KIAA0060; oscillin; zgc:110691 | |
| Mouse | |
| Protein G | |
| RUO | |
| 26384 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 237CT2.5.2 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O88958 | |
| GNPDA1 | |
| GNPDA1 recombinant protein. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction