missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNPDA1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97217JF549
This item is not returnable.
View return policy
Description
GNPDA1 Polyclonal antibody specifically detects GNPDA1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GNPDA1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| EC 3.5.99.6, GlcN6P deaminase 1, glucosamine-6-phosphate deaminase 1KIAA0060HLNGNPIGNPDA, glucosamine-6-phosphate isomerase, glucosamine-6-phosphate isomerase 1, GNP1, GNPDA 1, GPI, oscillin | |
| Produced in rabbits immunized with E. coli-derived Human GNPDA1 fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10007 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction