missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04642-100UL
This item is not returnable.
View return policy
Description
GNL2 Polyclonal specifically detects GNL2 in Mouse samples. It is validated for Western Blot.
Specifications
| GNL2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29889 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Autoantigen NGP-1, FLJ40906, guanine nucleotide binding protein-like 2 (nucleolar), HUMAUANTIG, Ngp-1, NGP1novel nucleolar guanosine 5'-triphosphate binding protein autoantigen, nucleolar G-protein gene 1, nucleolar GTPase, nucleolar GTP-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GNL2 (NP_037417.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction