missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GNL2 Polyclonal antibody specifically detects GNL2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | GNL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_037417.1 |
| Gene Alias | Autoantigen NGP-1, FLJ40906, guanine nucleotide binding protein-like 2 (nucleolar), HUMAUANTIG, Ngp-1, NGP1novel nucleolar guanosine 5'-triphosphate binding protein autoantigen, nucleolar G-protein gene 1, nucleolar GTPase, nucleolar GTP-binding protein 2 |
| Host Species | Mouse |
| Immunogen | GNL2 (NP_037417.1, 1 a.a. - 731 a.a.) full-length human protein. MVKPKYKGRSTINPSKASTNPDRVQGAGGQNMRDRATIRRLNMYRQKERRNSRGKIIKPLQYQSTVASGTVARVEPNIKWFGNTRVIKQSSLQKFQEEMDTVMKDPYKVVMKQSKLPMSLLHDRIRPHNLKVHILDTESFETTFGPKSQRKRPNLFASDMQSLIENAEMSTESYDQGKDRDLVTEDTGVRNEAQEEIYKKGQSKRIWGELYKVIDSSDVVVQVLDARDPMGTRSPHIETYLKKEKPWKHLIFVLNKCDLVPTWATKRWVAVLSQDYPTLAFHASLTNPFGKGAFIQLLRQFGKLHTDKKQISVGFIGYPNVGKSSVINTLRSKKVCNVAPIAGETKVWQYITLMRRIFLIDCPGVVYPSEDSETDIVLKGVVQVEKIKSPEDHIGAVLERAKPEYISKTYKIDSWENAEDFLEKLAFRTGKLLKGGEPDLQTVGKMVLNDWQRGRIPFFVKPPNAEPLVAPQLLPSSSLEVVPEAAQNNPGEEVTETAGEGSESIIKEETEENSHCDANTEMQQILTRVRQNFGKINVVPQFSGDDLVPVEVSDLEEELESFSDEEEEEQEQQRDDAEESSSEPEEENVGNDTKAVIKALDEKIAKYQKFLDKAKAKKFSAVRISKGLSEKIFAKPEEQRKTLEEDVDDRAPSKKGKKRKAQREEEQEHSNKAPRALTSKERRRAVRQQRPKKVGVRYYETHNVKNRNRNKKKTNDSEGQKHKRKKFRQKQ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?