missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GNL1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16812564
|
Proteintech
14078-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16802564
|
Proteintech
14078-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.Specifications
| GNL1 | |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P36915 | |
| 2794 | |
| GNL1 Fusion Protein Ag5203 | |
| Primary | |
| -20°C | |
| GNL1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GNL1, GTP binding protein HSR1, HSR1 | |
| GNL1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title