missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNL1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total GNL1 protein.
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
Specifications
| Antigen | GNL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GNL1 |
| Gene Accession No. | P36915, P36916, Q6MG06 |
| Gene Alias | G protein nucleolar 1 (putative); Gnal1; Gna-rs1; GNL1; GTP-binding protein HSR1; GTP-binding protein MMR1; guanine nucleotide binding protein, related sequence 1; guanine nucleotide binding protein-like 1; guanine nucleotide-binding protein-like 1; HSR1; HSR1 GTP-binding protein; Mmr1 |
| Gene Symbols | GNL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?