missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNGT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565357
This item is not returnable.
View return policy
Description
Immunogen sequence: EQLKKEVKNTR IPISKAGKEI KEYVEAQAGN DP Highest antigen sequence identity to the following orthologs - mouse 82%, rat 82%.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| GNGT2 | |
| Polyclonal | |
| Unconjugated | |
| GNGT2 | |
| AV096488; G gamma-C; G protein cone gamma 8 subunit; G protein subunit gamma transducin 2; gamma-T2 subunit; G-gamma-8; G-gamma-9; G-GAMMA-C; GNG8; GNG9; Gngt2; Gngt8; guanidine nucleotide binding protein, (G protein) , gamma transducing activity polypeptide 1; guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2; guanine nucleotide binding protein (G protein), gamma transducing polypeptide 2; guanine nucleotide binding protein gamma 9; guanine nucleotide binding protein gamma transducing activity polypeptide 2; guanine nucleotide binding protein, gamma transducing activity polypeptide 2; guanine nucleotide regulatory protein gamma c subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2; Tgamma c; transducin gamma subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2793 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14610 | |
| GNGT2 | |
| Recombinant Human GNGT2. Recombinant protein control fragment (Product #RP-106041). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction