missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141037
This item is not returnable.
View return policy
Description
GNG3 Polyclonal Antibody for Western Blot
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| GNG3 | |
| Polyclonal | |
| Unconjugated | |
| GNG3 | |
| [g]3; G protein subunit gamma 3; Gng3; Gngt3; G-protein gamma 3 subunit; guanine nucleotide binding protein (G protein), gamma 3; guanine nucleotide binding protein (G protein), gamma 3 subunit; guanine nucleotide binding protein, gamma 3; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3; guanine nucleotide-binding protein gamma-3 subunit; guanine nucleotide-binding regulatory protein gamma-3 subunit; NBP gamma-3; wu:fj56b02; zgc:92852 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114117, 14704, 2785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P63215, P63216 | |
| GNG3 | |
| Synthetic peptide within human GNG3 aa 25-75. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction