missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GNG13 Polyclonal antibody specifically detects GNG13 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | ELISA |
| Dilution | ELISA, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human GNG13 (Uniprot#: Q9P2W3; Met1-Cys64) |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?