missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNG11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100666
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GNG11.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| GNG11 | |
| Polyclonal | |
| Unconjugated | |
| GNG11 | |
| 0610037B21Rik; G protein gamma-11 subunit; G protein subunit gamma 11; GNG11; GNGT11; guanine nucleotide binding protein (G protein), gamma 11; guanine nucleotide binding protein gamma subunit 11; guanine nucleotide-binding protein G(I)/G(S)/G(O) gamma-11 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2791, 64199, 66066 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61952, P61953, P61954 | |
| GNG11 | |
| A synthesized peptide derived from human GNG11(Accession P61952), corresponding to amino acid residues M1-C36. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction