missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNF351, MedChemExpress
Description
GNF351 is a full aryl hydrocarbon receptor (AHR) antagonist. GNF351 competes with a photoaffinity AHR ligand for binding to the AHR with an IC50 of 62 nM. GNF351 is minimal toxicity in mouse or human keratinocytes.
Specifications
Specifications
| Chemical Name or Material | GNF351 |
| CAS | 1227634-69-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H25N7 |
| Quantity | 25 mg |
| Solubility Information | DMSO : ≥ 125 mg/mL (303.77 mM) |
| SMILES | CC1=CN=CC(C2=NC(NCCC3=CNC4=C3C=CC=C4)=C5N=CN(C(C)C)C5=N2)=C1 |
| Molecular Weight (g/mol) | 411.5 |
| Formula Weight | 411.5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?